Learn More
Enzo Life Sciences Mas 17 (1mg)
Supplier: Enzo Life Sciences BMLG4210001
Inactive analog of mastoparan (BML-G410). May be used as a negative control. Purity: ≥98%. Solubility: Soluble in Water. Long Term Storage: -20°C.