Learn More
Medchemexpress LLC HY-N1374 5mg Medchemexpress, Magnolin CAS:31008-18-1 Purity:>98%
Supplier: Medchemexpress LLC HYN13745MG
Medchemexpress, HY-N1374 5mg Magnolin CAS:31008-18-1 Magnolin, a major component of Magnolia flos (Shin-Yi), inhibits the Ras/ERKs/RSK2 signaling axis by targeting the active pocket of ERK1 and ERK2 with IC 50 s of 87 nM and 16.5 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.