Learn More
Electron Microscopy Sciences Magic Wand with Fine Top And Flat Tip, 6"
Supplier:  Electron Microscopy Sciences 6332301
Encompass_Preferred
Magic Wands are made from PEEK, a high performance plastic, with excellent chemical resistantance and withstands high temperatures (up to 300 degree C). Magic wand is ideal for use for electronic high-tech areas to laboratories, from preparation of samples to applying adhesives, using it as a stirring rod, or evendosing samples etc.
OAL: 6"
Catalog No. 50-322-31