Learn More
Accurate Chemical Lymphoprep, 4x250ml
Supplier: Accurate Chemical AN1001967
Lymphoprep, Isolation-Human Mononuclear Cells with Sodium diatrizoate (Ficoll-Isopaque) & polycaccharide. dens.=1.077g/mL, 280mOsm. 4x250 ml