Learn More
Electron Microscopy Sciences Low Heat Cautery Set
Supplier:  Electron Microscopy Sciences 7297610
Encompass_Preferred
The cautery comes with fully changeable tungsten carbide tips and batteries. Dual power: 1 AA battery for lower power and 2 AA batteries for higher power. Fully autoclavable. Stainless steel construction. Generates 300 to 350 deg C.
Catalog No. 50-298-35