Learn More
Electron Microscopy Sciences Applied Physics CeB6 Cathode Leica Lithography Philips
Supplier:  Electron Microscopy Sciences 80929P
Encompass_Preferred
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Philips base.
Type: CEBIX (CeB6)
Angle: 90°
Tips: 16 mm 
Catalog No. 50-949-871