Learn More
LaMotte™ Direct Reading Titrator
SDPSCI_ED
For applications prohibiting use of glass.
Supplier:  LaMotte™ 0377
Catalog No. S04523
Add to Cart
Description
Description
  • Chemically resistant plastic
  • One-piece plunger
  • No usage of rubber tips
  • Precision tip increases titration sensitivity
  • Direct replacement for LaMotte glass direct reading titrators
  • Available in all standard ranges
Specifications
Specifications
Direct reading titrator
Precision
Product Suggestions
Product Suggestions