Learn More
Medchemexpress LLC LAH5, 10 mg, CAS No. : 476169-12-7
Supplier: Medchemexpress LLC HYP1006810MG
LAH5 is an amphipathic cell-penetrating peptide. LAH5 effectively delivers CRISPR-Cas9 components into the nuclei of target cells by forming nanocomplexes with both RNP and RNP/HDR cargo.