Learn More
Volu Sol Lactophenol Cotton Blue, 1 oz / 30 mL, Volu-Sol™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Volu Sol VLC030
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Used for mounting medium and a staining agent used in the preparation of slides for microscopic examination of fungi. Fungal elements are stained intensely blue