Learn More
Bellco Glass, Inc. Blades and impeller assemblies are use for cell culture. Reduces shear in delicate cell culture. Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Blade dimension: 5 3/4 x 2". Shaft oal: 6 3/4".
Supplier: Bellco Glass, Inc. 196488052
Blades and impeller assemblies are use for cell culture. Reduces shear in delicate cell culture. Manufactured from 316l stainless steel with electropolish finish. Ra 32 finish or better inside and out. Blade dimension: 5 3/4 x 2". Shaft oal: 6 3/4".