Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ Kromasil C8 HPLC Column, 100 Å, 5 μm Particle Size
For small molecules and peptides.
$1077.00 - $1184.00
Specifications
Particle Size | 5 μm |
---|---|
Pore Size | 100 Å |
Stationary Phase | C8 (Octyl) |
Product Line | Kromasil |
Matrix | C8 (Octyl) Phase |
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | ||||
11-100-3541
|
MilliporeSigma™ Supelco™
K08670766 |
4.6 mm | 150 mm |
Each for $1,077.00
|
|
||||
11-102-0738
|
MilliporeSigma™ Supelco™
K08670369 |
4.6 mm | 250 mm |
Each for $1,184.00
|
|
||||
Description
Kromasil Classic 100 Å family of products is used to separate and purify molecules of up to about 10,000 Da.Specifications
5 μm | |
C8 (Octyl) | |
C8 (Octyl) Phase |
100 Å | |
Kromasil | |
Reversed Phase |
Kromasil is a registered trademark of EKA Chemicals AB
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title