Learn More
Alert:
This item is a non-standard voltage; please ensure you are ordering the correct product. Please contact customer service for assistance: 1-800-766-7000.
Description
Kinematic viscosity is of primary importance in the design and selection of a wide range of petroleum products.
- Accommodates six round 2 in. dia. (51mm) viscometer holders
- Test temperatures of up to 150°C (302°F) can be selected
- Temperature limit control permits operator to select an overtemperature cutoff point to protect against accidental overheating
- Control unit includes immersion heater, circulating stirrer and temperature probe
- Bath Medium: water or white technical oil (sold separately)
- Thermometers sold separately
Compliance
Complies with ASTM D445, D6074, D6158; IP 71; ISO 3104; DIN 51550;FTM 791-305; and NF T 60-100 specifications.
Specifications
5.8 gal. | |
13.5 x 20 in. (34.6 x 50.8 cm) | |
12 in. (31cm) |
22 L | |
KV1000 | |
25 lb. |