Description
- Includes 10 sets of glassware, heater, and pre-cut tubing.
Specifications
10 sets of glassware, heater, and pre-cut tubing | |
Cyanide Distillation System | |
USP Type I and ASTM E438, Type I, Class A |
Designed to support the semi-automatic colorimetric methods for drinking, ground, surface and saline waters as well as domestic and industrial wastes. | |
Borosilicate Glass |