Learn More
DWK Life Sciences Kimble™ KIMAX™ Thistle-Tube Funnel
SCI_ED
Highly functional.
Supplier:  DWK Life Sciences 4618865300/EMD
Catalog No. S30814
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
Description
Description
This cylindrical-shaped tube offers two different stem lengths. Its thistle-top style makes this funnel useful for adding reagents to reaction vessels.
Specifications
Specifications
6.5mm
Used for adding reagents to reaction vessel
Thistle Tube Funnel
Borosilicate Glass
300mm
Product Suggestions
Product Suggestions