Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Marking spot is durable fused-on white enamel
Supplier:  DWK Life Sciences 4504220150
 View more versions of this product
Catalog No. 14-956G
Add to Cart
Description
Description
Marking spot is durable fused-on white enamel
  • Kimax KG-33 glass
  • With sturdy lip
  • Uniform wall thickness for maximum heat transfer, chemical resistance
Specifications
Specifications
ASTM E982, Type I
36 mL
Borosilicate Glass
20 mm
Plain Beaded Rim
150 mm
Marking Spot
Reusable Test Tube
Product Suggestions
Product Suggestions