Learn More
DWK Life Sciences Kimble™ KIMAX™ Reusable Test Tubes with Beaded Rim and Marking Spot
Marking spot is durable fused-on white enamel
Supplier:  DWK Life Sciences 450421275
 View more versions of this product
Catalog No. 14-956B
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Marking spot is durable fused-on white enamel
  • Kimax KG-33 glass
  • With sturdy lip
  • Uniform wall thickness for maximum heat transfer, chemical resistance
Specifications
Specifications
ASTM E982, Type I
Plain Beaded Rim
75 mm
Marking Spot
12 mm
6 mL
Borosilicate Glass
Product Suggestions
Product Suggestions