Learn More
Kemtec™ Seawater Analysis Kit
Students analyze chloride content of seawater samples.
Supplier: Aqua Phoenix Scientific 4102
Includes: Silver nitrate, potassium chromate indicator, Atlantic Ocean water, Pacific Ocean water, Gulf of Mexico water, instructor manual, SDS, assessment manual and student data sheets
Requires : Goggles and 25mL burets.
Description
- Students use volumetric titration technique and analyze samples from the Atlantic Ocean, Pacific Ocean and Gulf of Mexico
- Water from local sources can also be analyzed
- Constant ratio of dissolved substances in sea water allows concentration of salt and other elements to be calculated from chlorine concentration
- Students use their lab data to compare and contrast climates and factors affecting salinity of Atlantic and Pacific Oceans and the Gulf of Mexico
- Investigate how earth is affected if climate change continues to disrupt the ocean conveyor belt system
- Grades 10 to 12
- Class size: 24 students
Specifications
Seawater Analysis Kit | |
Analyze chloride content of seawater samples | |
Instructor manual, msds, assessment manual, student data sheets, special needs, silver nitrate solution, potassium chromate indicator solution, atlantic ocean water, pacific ocean water, gulf of mexico water |