Learn More
Dynalon™ Kartell Tray for Sample Bottles
SCI_ED
For holding sample bottles with base diameter not greater than 22mm or less; e.g. Bijou bottles, specimen vials. Dynalon™ Kartell Tray for Sample Bottles are constructed of sturdy PVC.
Supplier:  Dynalon™ 259214
Catalog No. S37260
Add to Cart
Description
Description
  • Ideal for 7mL (1/4oz) Bijou bottles or specimen tubes
  • Consists of 9 rows
  • Each row will hold 12 to 15 small sample vials
  • Makes filling stable, organized and easy
Specifications
Specifications
PVC
12 to 15 vials
350 x 300 x 40 mm
Product Suggestions
Product Suggestions