Learn More
Thermo Scientific ION PAC COLUMN AS11-HC
Supplier: Thermo Scientific 052960
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
IonPac AS11-HC Analytical Column; 0 to 14 pH; 4 x 250mm; Bead dia.: 9um; Pore Size: 2000 angstrom; Capacity: 290ueq; 4000psi(OP); Use the ICE-AS6 ion-exclusion columns for organic acid analysis in high-ionic-strength samples
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)