Learn More
BioMed Diagnostics An enriched dermatophyte medium used in the detection of dermatophytes. | Detects: Trichophyton Mentagrophytes, T. rubrum, T. tonsurans, Microsporum gypseum. | Available in: 20 pack
Supplier:  BioMed Diagnostics 12063002
Encompass_Preferred
InTray DM is an enriched dermatophyte medium used in the detection of dermatophytes from clinical specimens. InTray DM is formulated to produce a red color in the presence of growing dermatophytes. The medium is formulated to produce distinctive colony growth with typical identifying characteristics both macro and microscopically. The medium inhibits most Gram-positive and Gram-negative bacteria, yeast and saprophytic fungi. | Detects: Trichophyton Mentagrophytes, T. rubrum, T. tonsurans, and Microsporum gypseum. | Time to Result: Incubate inoculated trays for up to 14 days at 18-25C, in the dark. Observe the trays daily for color changes, through the clear viewing window. | Available in: 20 pack
Catalog No. NC2582845