Learn More
Rotronic Instrument Corp HYGROPALM HP32 W/O PROBE
SDP
Supplier:  Rotronic Instrument Corp HP32A
Encompass_Preferred
HYGROPALM HP32 (probe sold separately)Package content• HygroPalm HP32 (without probe)• Soft case• Micro-USB cable (computer connection and battery charging)• HW4 LITE license key (max. 3 instruments connected)Benefits• Relative humidity, temperature and psychrometric calculations• Graphical display for graphs and up to 4 values• Freely settable visual and audible alarms on every value• 64,000 data points memory with programmablelogging interval• Long-life rechargeable battery (up to 48 hours at a logging intervalof 30 seconds)Applications• Field inspection and spot checks in HVAC• Building management systems• Agriculture and food industry• Pharmaceutical industry
Catalog No. NC2162323