Learn More
Azzota Corp Hydrogen Peroxide Sterilizer, OA: 80m/s, OV:: 150ml/h, Max. WV: 150 m3, C: 8ml/m3, 300W, 110V, 50Hz-60Hz,500ml
Supplier:  Azzota Corp AZIHP
Encompass_Preferred
Request Bulk Order
Hydrogen Peroxide Sterilizer, OA: 80m/s, OV:: 150ml/h, Max. WV: 150 m3, C: 8ml/m3, 300W, 110V, 50Hz-60Hz,500ml.
Catalog No. NC2329948