Learn More
Research Products International Corp Hybrilinker Hybridization Oven and UV Crosslinker
SDP
Supplier:  Research Products International Corp 950031
Encompass_Preferred
Request Bulk Order
The ideal solution for overcrowded laboratory bench space. The Hybrilinker enables all hybridization protocols, washing and UV crosslinking in one compact size unit.
Hybridization Oven has accurate temperature control from ambient to +100°C and variable rotation speed of 0-18 rpm. Accepts up to twenty 35 x 150 mm or ten 35 x 300 mm bottles. Optional interchangeable rocking platform available. Crosslinker provides a uniform 254 nm UV exposure monitored by a microprocessor for accurate and safe crosslinking. The hybridization oven and crosslinker features separate power supplies for independent operation.
Catalog No. 50-197-6806
Add to cart
missing translation for 'provideContentCorrection'
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
missing translation for 'productTitle'