Learn More
Caron Products And Services Humidified Incubator, 10 cu. ft., Temp & RH
Supplier:  Caron Products And Services 7003102
Encompass_Preferred
Request Bulk Order
10 ft3 (283 L) Humidified Incubator offers temperature (10C above ambient to 70C) and humidity control (ambient to 98% RH). Ideal for a wide range of applications. Delivers dependable performance and reliable results. Includes earth-friendly and customer convenient features. 208/230V, 60 Hz.
Catalog No. NC2676696