Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human IL-1 beta/IL-1F2 Quantikine ELISA Kit

Human IL-1 beta/IL-1F2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

$736.60 - $4199.20

Specifications

Assay Range 3.90 - 250pg/mL
Assay Sensitivity 1pg/mL
Conjugate HRP
Cross Reactivity <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Product Type Solid Phase Sandwich ELISA
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
DLB50
View Documents
R&D Systems™
DLB50
1 Plate
Each for $736.60
Add to Cart
 
SLB50
View Documents
R&D Systems™
SLB50
6 Plates
Each for $4,199.20
Add to Cart
 
Description

Description

The Human IL-1 beta/IL-1F2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-1 beta/IL-1F2 in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul), citrate plasma (200 ul).
Specifications

Specifications

3.90 - 250pg/mL
HRP
Solid Phase Sandwich ELISA
Natural and Recombinant Human IL-1 beta
ELISA
Cell Culture Supernates (200uL),Serum (200uL),EDTA Plasma (200uL),Heparin Plasma (200uL),Citrate Plasma (200uL)
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1 b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta
1pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum
Human
3553
Store the unopened product at 2° to 8°C. Do not use past expiration date.
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit