Learn More
Electron Microscopy Sciences EMS Holey Carbon Film 50 µm holes on 200 mesh Nickel
Supplier:  Electron Microscopy Sciences HC200NI
Encompass_Preferred
A thin film of carbon with round holes. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
Holes: 50 µm
Catalog No. 50-244-44