Learn More
DWK Life Sciences Wheaton™ Crimping Jaw Set for High Performance Crimping Tool
Interchangeable jaws allow users to choose a jaw that best fits their needs.
Supplier:  DWK Life Sciences W225762
 View more versions of this product
Catalog No. 02-915-020
Add to Cart
Description
Description
Available as crimper and decappers which fit standard lab vials seals with aluminum or two-part aluminum/steel caps
Specifications
Specifications
Decapping
High performance crimping tool
13 mm
Product Suggestions
Product Suggestions