Learn More
Thermo Scientific™ Herasafe™ 2030i CONFIGURED-NSF Biosafety Cabinet PROMO
Herasafe 2030i incorporates SmartFlow Plus, DAVe Plus and Smart UV technologies for ultimate protection and graphical user interface for ease of operation. Unit includes inbuilt Wi-Fi capability and is ready to be 'connected'.
$13790.00 - $16870.00
Specifications
Certifications/Compliance | NSF/ANSI 49 |
---|---|
Chamber Material | Stainless Steel (316) |
Product Type | Biosafety Cabinet |
Model | Herasafe 2030i |
Sash Opening (Metric) | 25.4 cm |
Catalog Number | Mfr. No. | Dimensions (L x W x H) Exterior | Outlets | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Dimensions (L x W x H) Exterior | Outlets | Price | Quantity & Availability | ||||
09-034-250PM
|
Thermo Scientific™
51032334TU |
80 x 130 x 154 cm (31.5 x 51.2 x 60.5 in.) | 2 Rear Wall |
Each for $13,790.00
|
|
||||
09-034-251PM
|
Thermo Scientific™
51032335TU |
80 x 190 x 154 cm (31.5 x 74.8 x 60.5 in.) | 4 Rear Wall |
Each for $16,870.00
|
|
||||
Description
Breakthrough innovations and unprecedented features make the Herasafe 2030i BSC Thermo Scientific™'s most technologically advanced and user-friendly Class II, Type A2 cabinet available. (Stand sold separately)
Smart protection:
- Thermo Scientific™ SmartFlow™ Plus Technology - Autonomous safety features maintain proper air flow and monitor critical conditions in real time without user interaction.
- Self-adjusting germicidal UV light intensity - Unique cross beam UV system has exclusive self-adjusting exposure capability to enable reproducible germicidal effectiveness with every use
- 316L stainless steel interior - robust and resistant to scratches and chemicals
Simplified operation and ergonomics:
- Touchscreen interface
- SmartClean Plus window design – Easily access the interior of the BSC with the motorized front sash which automatically stops at the set working height. The sash can be further opened electronically or the whole front panel hinges open for thorough cleaning access.
- Bright work area with glass side walls and LED lights
- Ergonomic Arm Rests (option)
- SmartPort convenience - Easily pass tubing and wiring through sidewall to accommodate many applications needs.
Information management:
- Touchscreen and connectivity (available in select countries) – Built-in WiFi system is ready for cloud-based capabilities like remote equipment monitoring, secure data storage, collaboration tools and more. Digital Airflow Verification (DAVe) alerts, system self-checks and performance status are provided on the full-color touchscreen.
Energy efficiency:
- LED interior illumination
- Dual DC motors
- Standby mode
Certifications:
- For North America, cabinets are NSF-49 certified (4 ft. and 6 ft. models)
- For Europe, the cabinets are certified according to EN12469 (all 4 sizes). The 1.2 m model has been certified to EN 12469 with the NF mark by LNE as well.
Available sizes:
- North America 4 ft. and 6 ft. models; other countries: 0.9 m, 1.2 m, 1.5 m and 1.8 m
For cell and gene therapy applications, we offer the Herasafe 2030i Biological Safety Cabinet CTS (Cell Therapy Systems) Series which consists of the product, a comprehensive documentation package including certifications and protocols, as well as compliance services.
Previous models: Herasafe KS Note: Please order stand separately and indicate any additional requirements/ options you need when you 'Request a quote.'
Specifications
NSF/ANSI 49 | |
Biosafety Cabinet | |
25.4 cm | |
0.99995 | |
Energy Efficient | |
2 Ports/Side with Additional Smart Ports | |
60 Hz |
Stainless Steel (316) | |
Herasafe 2030i | |
7 in. GUI | |
H14 HEPA | |
LED | |
10 in. | |
120 V |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.