Learn More
Volu Sol Hematology Buffer (pH 6.8), 32 oz / 1 L, Volu-Sol™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Volu Sol VWB032
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Phosphate Buffer, intended for use as a buffer in various Romanowsky type staining procedures (Wright, Wright-Giemsa, Giemsa, May Grunwald, Jenner, and Leishman)