Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Blunt Tip, PEEK
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Supplier:  MilliporeSigma™ Supelco™ 26710U
Catalog No. 11-100-9600
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Specifications
Specifications
PEEK (Tip)
2 in.
6/Pk.
51 mm
Hamilton RN Syringes
16 G
Blunt Tip
250 μL to 10 mL
3
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Hamilton is a registered trademark of Hamilton Co.