Learn More
Creative Biomart Recombinant CoV-2 N-Mosaic Antigen, His-tagged
Supplier:  Creative Biomart H1V1303
Encompass_Preferred
The E. coli derived recombinant protein contains the Coronavirus 2019 full length nuclepocapsid Mosaic immunodominant regions [full length N-antigen], fused to 6 His tag at C-terminal.
Catalog No. 50-208-4681
Due to product restrictions, please to purchase or view availability for this product.