Learn More
Waters Corp GTxResolve Premier SEC 1000Å, 3 μm, 4.6 x 300 mm
Supplier: Waters Corp 186010736
The GTxResolve Premier SEC 1000Å, 3 μm, 4.6 x 300 mm Column is manufactured with low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles to support high recovery analyses of 200 to 800Å diameter GTx drugs, such as mRNA, LNPs, and viral vectors. This column technology provides long column lifetimes, low secondary interactions, and improved MALS sensitivity. Facilitate the development of platform multiattribute SEC assays for mRNA integrity and LNP formulations analyses as well as improved detection of ejected transgenes and aggregates related to viral vectors
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.