Learn More
Med Vet International GOWN SURGEON N/S DISP LARGE
Supplier:  Med Vet International AHSGNSL
Encompass_Preferred
These gowns are made of a non-woven material that helps provide a barrier to blood, body fluids and pathogens. They offer a high level of protection without compromising comfort. They are cut wider through the chest and sleeves for a roomier fit and feature a velcro-style neck closure that allows the neckline to easily adjust to a wide variety of people. In addition, the velcro-style closure helps prevent gapping in the front of the gown to further protect against fluids. The gowns are very low linting, minimizing the risk of introducing particles into the O.R.Level 2
Catalog No. 50-252-2458
Due to product restrictions, please to purchase or view availability for this product.