Learn More
Pfanstiehl Inc L-Histidine, High Purity, Low Endotoxin, Low Metals, USP, EP, JP, ChP (H-116) - 1kg pack – GMP, Multicompendial - CAS: 71-00-1
Supplier: Pfanstiehl Inc H1161KG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Histidine is commonly used in biotherapeutic formulations as well as bioprocessing. Histidine has been shown to protect monoclonal antibodies in both the liquid and lyophilized states against heat stress and is becoming increasingly common in formulations of protein therapeutics. Histidine is the only amino acid with pH 7.4 within its buffering range, leaving histidine as the best amino acid buffer under physiological conditions. Pfanstiehl High Purity / Low Endotoxin-Low Metal Histidine establishes a new quality standard for amino acids, designed specifically for Biopharmaceutical formulation and bioprocessing. Pfanstiehl Brand Histidine is available as USP (United States Pharmacopoeia), EP (European Pharmacopoeia), JP (Japanese Pharmacopoeia) and ChP (Chinese Pharmacopoeia).
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.