Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eisco™ Glass Stirring Rods, Rounded Ends, Pack of 10
Glass Stirring Rods are made from high quality borosilicate glass and feature two rounded ends.
Supplier: Eisco™ CH0754
Description
Warranty and Services
1 year limited manufacturer warranty
Specifications
Sphere | |
Stiring | |
7 mm | |
20 cm |
10 | |
Yes | |
Borosilicate Glass | |
Clear |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction