Learn More
Enzo Life Sciences Ginkgolide B (10mg). CAS: 15291-77-7
SDP
Supplier:  Enzo Life Sciences BMLL1350010
Encompass_Preferred
Most potent member of the ginkgolide family of PAF antagonists. Ginkgolide B accelerates PAF hydrolysis by promoting PAF-AH I α2 homodimer activity. Inhibits PAF-induced PMNL chemotaxis (IC50=490 nM). Orally effective against PAF-induced bronchospasm. Alternative name: BN 52021. Purity: ≥90% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (2mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-1137