Learn More
Gibco™ Fetal Bovine Serum, certified, United States
Important Update: We've recently introduced Premium Plus Fetal Bovine Serum (FBS) specification-based products. These improved products are alternatives to our Certified US origin, New Zealand origin, and Qualified Australia orgin SKUs. Product stability, performance, manufacturing, storage, and transport processes are unchanged.
$98.65 - $1528.65
Specifications
Content And Storage | Storage conditions: ≤-10°C Shipping conditions: Frozen |
---|---|
Country of Origin | United States |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Serum Treatment | Standard (Sterile-filtered) |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
16-000-044
|
Gibco™
16000044 |
500 mL |
Each
for $787.65
Save 6% Reg : |
|
|||||
16-000-069
|
Gibco™
16000069 |
1000 mL |
Each
for $1,528.65
Save 6% Reg : |
|
|||||
A3160401
|
Gibco™
A3160401 |
50 mL |
Each
for $98.65
Save 5% Reg : |
|
|||||
A3160402
|
Gibco™
A3160402 |
10 x 50 mL |
Each
for $929.65
Save 6% Reg : |
|
|||||
Description
- Used for broad range of cell types, especially sensitive cell lines
- With lowest endotoxin and hemoglobin levels
- Endotoxin level: ≤ 5 EU/mL
- Hemoglobin level: ≤ 10mg/dL
- Origin: United States
- ISO 13485 certified, processed in FDA registered facilities
- Labeled for in-vitro diagnostics (compliant with highest level of USP sterility testing)
- Triple filtered at 0.1μm
- Gibco™ bottle is easier to use in hood, reduces risk of contamination and helps to perform cell culture more consistently
- Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
Cell Culture, Drosophila S2 Cell Culture, High Five™ Cell Culture, Insect Cell Culture, Mammalian Cell Culture, Sf9 and Sf21 Cell Culture
Order Info
Shipping Condition: Dry Ice
Specifications
Storage conditions: ≤-10°C Shipping conditions: Frozen |
|
Liquid | |
Standard (Sterile-filtered) | |
≤5 EU/mL | |
Certified |
United States | |
Fetal Bovine Serum | |
Sterile | |
≤15 mg/dL | |
Frozen |
For In Vitro Diagnostic Use.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.