Learn More
Giant Eye in Bony Orbit
SDPSCI_ED
Supplier:  Denoyer Geppert Company 018200
Catalog No. S171153
Add to Cart
Description
Description
Specifications
Specifications
Model, Eye
Model re-creates how the eye moves in the socket to provide a better understanding of its relationship with major blood vessels, muscles and nerves
23 cm
12 in.
Vinyl
Giant
25 cm
12 x 10 x 9 in. (30 x 25 x 23 cm)
9 in.
Horizontally sectioned sclera and vitreous humor, cornea and iris, optic nerve, lens, superior and lateral removable rectus muscles, manual identifying 77 structures
30 cm
Bony Orbit
10 in.
Product Suggestions
Product Suggestions