Learn More
Santa Cruz Biotechnology Gel Incubation Tray, Blue, 100/pk
Supplier:  Santa Cruz Biotechnology SC201755
Encompass_Preferred
Gel incubation tray is made of polystyrene. Gel incubation tray is commonly used in blotting applications such as incubating and staining gels and membranes.
Catalog No. NC1863513