Learn More
Carolina™ Formalin Preserved Sheep Brain, Dura Mater Removed
Carolina™ Formalin Preserved Sheep Brain with the Dura Mater Removed is used to study mammalian brain structure.
Supplier: Carolina™ 228700
This item is not returnable.
View return policy
Description
- Similar to human brain
- Sheep brain with dura mater removed and optic chiasma intact
- Shipped in Carosafe™, Carolina's proprietary, formaldehyde-free holding fluid, designed to minimize odor and formaldehyde exposure
- Plain: No color injection
Specifications
Preserved Specimen | |
Plain | |
Pail | |
Non-sterile |
Formalin | |
1/Pk. | |
Not Autoclavable | |
No |