Learn More
STA PHARMACEUTICAL US LLC WuXi TIDES Fmoc-L-Phe(3,5-DiCl)-OH | 1260605-95-5, 100GR
Supplier:  STA PHARMACEUTICAL US LLC STAAA10132100GM
Encompass
Request Bulk Order
WuXi TIDES Fmoc-L-Phe(3,5-DiCl)-OH, 1260605-95-5 | (S)-2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-(3,5-dichlorophenyl)propanoic acid | O=C(N[C@@H](CC1=CC(Cl)=CC(Cl)=C1)C(=O)O)OCC2C3=CC=CC=C3C4=C2C=CC=C4 | M.F. C24H19Cl2NO4, M.W. 456.32
Catalog No. 50-235-0233