Learn More
Perkin Elmer US LLC Flowcell Assembly, 3mm x 1.7µL , for Series 200/785A and ABI 759A UV/VIS Detector, Prep
Supplier:  Perkin Elmer US LLC 29000545
Encompass_Preferred
Flowcell Assembly, 3mm x 1.7µL , for Series 200/785A and ABI 759A UV/VIS Detector, Prep
Catalog No. 50-211-3284