Learn More
MSC FLIR MR160 32 to 122°F Operating Temp, Moisture Meter Color LCD Display, Accurate to ±5%, Includes MR02 Pin Probe, Quick Start Guide, International Charger, Warranty Card, Brochure Card
Supplier:  MSC 59235333
Encompass
FLIR MR160 32 to 122°F Operating Temp, Moisture Meter Color LCD Display, Accurate to ±5%, Includes MR02 Pin Probe, Quick Start Guide, International Charger, Warranty Card, Brochure Card
Catalog No. NC1824641