Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Healthcare™ Sure-Vue™ Color Staph ID Latex Test Kit
The Sure-Vue Color Staph is a rapid, color enhanced latex agglutination test to detect clumping factor and/or Protein-A, two characteristics associated with Staphylococcus aureus colonies obtained from culture.
Specifications
CE Marker | Self-declared |
---|---|
Clia Complexity | Moderate (Serum, Plasma or Whole Blood), Waived (Other) |
Detectable Analytes | S. aureus |
Type | Sure-Vue™ Color Staph ID Latex Test Kit |
Disposable | Single-use |
Includes: Latex particle reagent, positive and negative controls, control latex reagent, disposable white slide cards (8-well), disposable mixing sticks, and package insert
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
23-046-500
|
150 Tests |
Pack of 150 for $365.00
|
N/A | ||||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-046-501
|
300 Tests |
Pack of 300 for $635.00
|
N/A
|
||||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- CLIA Complexity: Non-Waived
- CPT Code: 87077
- Sensitivity: 99%
- Specificity: 100%
- Latex agglutination slide test for the qualitative detection of Staphylococcus aureus from primary cultures
- Produces visible red agglutination in a blue background when results are positive
- Results within 30 seconds
- Unused reagents can safely be used up to expiration date stamped on the box with prompt recapping and refrigeration
- For in vitro diagnostic use
Specifications
Self-declared | |
S. aureus | |
Single-use | |
Ready-to-Use (RTU) |
Moderate (Serum, Plasma or Whole Blood), Waived (Other) | |
Sure-Vue™ Color Staph ID Latex Test Kit | |
Latex particle reagent, positive and negative control, control Latex reagent, disposable white slide cards (8-well) and disposable mixing sticks |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title