Learn More
Kamiya Biomedical Fibrinogen Control, L1: 5 x 1mL, L2: 5 x 1mL RX
Supplier: Kamiya Biomedical K63C10M
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
K-ASSAY Fibrinogen Control. Level 1: 5 x 1 mL (lyophilized), Level 2: 5 x 1 mL (lyophilized). The K-ASSAY Fibrinogen Control is intended for use in the quality control of fibrinogen assays, for example the K-ASSAY Fibrinogen immunoturbidimetric assay (Cat. No. KAI-135). FOR IN VITRO DIAGNOSTIC USE.