Learn More
LabStrong Corp FI-STREEM HEATING ELEMENT 208V
Supplier: LabStrong Corp 562X62LS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
LabStrong Fi-Streem part number 562X62LS compares to Fi-Streem part number 562X62. The heating element (208V, 1.5Kw) is a replacement part for Fi-Streem Water Purification Stills. The heating element includes the plastic element union and heater sheath.