Learn More
Perkin Elmer US LLC Fast Stabilization PTFE Rotor for High-Flow 7-Port FS Valve
Supplier: Perkin Elmer US LLC N8145264
This PTFE Rotor is for use with the high-flow 7-port Fast Stabilization (FS) valve in SC-FAST Autosampler Systems. The complete valve features 7 ports and combines the benefits of high-flow with fast stabilization times. The FS valve has the fastest stabilization and rinse out (<5 s).