Learn More
Restek™ EZ Twist Top Injection Port Kits
Change inlet liners faster and easier, for Agilent 5890/6850/6890/7820 GCs
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | For Use With (Equipment) | Price | Quantity & Availability | ||||
06-611-234
|
Restek™
22725 |
Injection Port Kit | Agilent 5890 |
Each for $936.00
|
N/A | ||||
06-611-235
|
Restek™
22726 |
Siltek™ Injection Port Kit | Agilent 5890 |
Each for $1,066.00
|
N/A | ||||
06-611-230
|
Restek™
22721 |
Injection Port Kit | Agilent 6850/6890/7820 |
Each for $1,015.00
|
N/A | ||||
06-611-231
|
Restek™
22722 |
Siltek™ Injection Port Kit | Agilent 6850/6890/7820 |
Each for $1,182.00
|
N/A | ||||
06-611-242
|
Restek™
22735 |
Injection Port Kit | Agilent 6850/6890/7820 (Large Canister Type Filter) |
Each for $1,182.00
|
N/A | ||||
06-611-243
|
Restek™
22736 |
Siltek™ Injection Port Kit | Agilent 6850/6890/7820 (Large Canister Type Filter) |
Each for $1,451.00
|
N/A | ||||
Description
- Gas lines are attached to shell weldment bottom to help eliminate broken gas lines
- Gas lines do not interfere with routine maintenance
- Weldment Removal Tool allows for quick removal of the hot weldment without fingers ever touching hot surfaces
- Weldment stays secure in the tool for reattachment
Specifications
EZ Twist Top |