Learn More
Electron Microscopy Sciences EMS 9000 Microwave Oven
Supplier:  Electron Microscopy Sciences 97030
Encompass_Preferred
Request Bulk Order
The EMS 9000 Laboratory Microwave Oven represents a state-of-the-art solution for performing many of the steps in sample preparation prior to either light or electron microscopy. Procedures such as fixation, staining, dehydration, decalcification, impregnation, polymerization as well as immunohisto and cytochemistry can be done with ease in the EMS 9000.
The EMS 9000 offers a significant reduction in processing times while obtaining improved results.
The unit features 825 watt nominal output with variable wattage, bubble manifold for 5 tubes, forced exhaust system with fail safe interlock, adjustable duty cycle-one second and greater for very precise process control, magnetic pre-warming, right side closet, three different times modes, multple safety interlocks, visual and aural warnings on errors and malfunctions, all controls automated, and flexible temperature probe.
Catalog No. 50-290-50