Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CDS Empore™ 96-Well Extraction Disk Plates
Formerly 3M™ Empore™ Solid Phase Extraction Plates: 6015SD, 6014SD, 6030SD. High-performance 96-Well Disk Plates for high-throughput SPE.
Supplier: CDS 70200739814EA
This item is not returnable.
View return policy
Description
- Process 96 samples in 8 x 12 plate formats compatible with liquid handling technologies and injection systems
- Samples can be extracted in approximately one hour or less
- Wells contain patented Empore standard-density particle-loaded membrane for efficient sample extraction followed by elution in 100μL with mobile phase or mobile-phase compatible solutions
- Ideal for sample preparation prior to LC/MS and other high-throughput applications
- C2, C8, or C18 silica sorbents for nonpolar interactions with analytes from complex samples
- Universal resin (UR) sorbent plates contain polar functioning groups to promote a much more generic approach with high retention of acidic, neutral, and base analytes
- Use with CDS Empore Vacuum Manifold Assembly when extracting multiple samples
Specifications
10 mg | |
Solid Phase Extraction, LC/MS | |
C8-SD | |
1.2 mL | |
Polypropylene | |
Universal | |
0.2 in. |
Solid Phase Extraction, LC/MS | |
40 to 60 μm | |
Extraction Disk Plate | |
1.2 mL | |
12/Cs. | |
12.5 cm | |
0.5 cm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction